missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum selenide, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
It is used as a precursor to hydrogen selenide
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Brown |
Physical Form | Pieces |
Quantity | 25 g |
UN Number | UN3283 |
Merck Index | 14,361 |
Solubility Information | Decomposes to Al203 and SeO2 in water |
Formula Weight | 290.84 |
Percent Purity | 99% |
Odor | Odorless |
Sensitivity | Moisture sensitive |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'