missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets
Designed for use in thin layer chromatography. Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets include aluminum oxide layers and a fluorescent indicator at short-wave UV (254nm). Activating the layers before use is recommended.
Brand: Macherey-Nagel™ 818024
This item is not returnable.
View return policy
Description
ALUGRAM™ Alox N/UV254 are aluminum sheets with aluminum oxide layers for use in TLC. They feature an inert organic binder and a manganese-activated zinc silicate indicator.
- Recommended application: terpenes, alkaloids, steroids, aliphatic and aromatic compounds
- Activate the aluminum oxide layers before use by heating 10 minutes at 120™C
Specifications
Aluminum Sheets Alugram Alox N | |
50 Pk. | |
UV254 | |
∼200 m2/g | |
5 x 20 cm |
60 Å | |
TLC Plate | |
Terpenes, Alkaloids, Steroids, Aliphatic and Aromatic Compounds | |
0.2 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction