Learn More
Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets
Designed for use in thin layer chromatography. Macherey-Nagel™ ALUGRAM™ Alox N/UV254 Aluminum Sheets include aluminum oxide layers and a fluorescent indicator at short-wave UV (254nm). Activating the layers before use is recommended.
Brand:  Macherey-Nagel™ 818023
125.63 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.
 View more versions of this product
Product Code. 11386474
  • 157.00 € / Pack of 25
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
ALUGRAM™ Alox N/UV254 are aluminum sheets with aluminum oxide layers for use in TLC. They feature an inert organic binder and a manganese-activated zinc silicate indicator.
  • Recommended application: terpenes, alkaloids, steroids, aliphatic and aromatic compounds
  • Activate the aluminum oxide layers before use by heating 10 minutes at 120™C
Specifications
Specifications
Aluminum Sheets Alugram Alox N
25 Pk.
UV254
∼200 m2/g
20 x 20 cm
60 Å
TLC Plate
Terpenes, Alkaloids, Steroids, Aliphatic and Aromatic Compounds
0.2 mm