missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

Brand:  Macherey-Nagel™ 818129

158.44 EUR valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 12820951

  • 175.00 € / Pack of 200
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Separate non-volatile mixtures with unmodified standard silica layers for TLC (Thin-Layer Chromatography). Standard SIL G Silica Layers on Alugram™ Aluminum Sheets UV254 come in a variety of sizes.

Alugram™ Aluminum Sheets for TLC
  • Pre-coated ready to use plates
  • Broad range of plate sizes, surface coatings and backings
Absorbent
  • Silica 60
  • specific surface (BET) ∼ 500 m2/g
  • Mean pore size 60 Å
  • Specific pore volume 0.75 mL/g,
  • Particle size 5–17 μm
  • Standard grade
Thickness
  • For analytical plates 0.25 mm
  • For preparative plates 0.5 and 1 mm
  • For 2 mm preparative layers a slightly coarser material is used.
Inicators
  • Manganese activated zinc silicate with green fluorescence for short-wave UV (254 nm)
  • Special inorganic fluorescent pigment with blue fluorescence for long-wave UV (366 nm)
Binders
  • Highly polymeric products, which are stable in almost all organic solvents and resistant towards aggressive visualisation reagents
Specifications

Specifications

Aluminum Sheets Alugram SIL G
60 Å
TLC Plate
0.75 mL/g
0.2 mm
5 to 17 μm
200 Pk.
UV254
∼500 m2/g
2.5 x 7.5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.