missing translation for 'onlineSavingsMsg'
Learn More

Aluminum shot, nominally 9.5mm (0.375in) flattened/spherical shape, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 15494925
Click to view available options
Quantity:
500 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Spheroid shape pellet 3/8 in. ±1/4 in.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Specifications

Color Silver
Appearance Solid in various forms
Assay Percent Range 99.99% (Metals basis)
Quantity 500 g
IUPAC Name aluminum
Odor Odorless
Form Pellet
Chemical Name or Material Aluminum

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'