missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11314388
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 11145-30-5
Molecular Formula AlSi
Molecular Weight (g/mol) 55.067
MDL Number MFCD00143595
InChI Key CSDREXVUYHZDNP-UHFFFAOYSA-N
Synonym silicon aluminum, aluminum-silicon alloy, al.si, aluminum silicon slug, aluminum silicon, aluminum-silane 1/1, aluminum silicon powder,-325 mesh, aluminum silicon slug, 6.35mm 0.25in dia x 6.35mm 0.25in length, aluminum silicon slug, 12.7mm 0.5in dia x 12.7mm 0.5in length, puratronic, aluminum silicon slug, 6.35mm 0.25in dia x 12.7mm 0.5in length, puratronic
PubChem CID 9793685
IUPAC Name aluminum;silicon
SMILES [Al].[Si]

Specifications

Avg. Mol. Wt. or Mol. Wt. Range Al:Si; 99:1 wt%
Quantity 250 g
Percent Purity 99.99%
Odor Odorless
Form Slug
Assay (metals basis)
Chemical Name or Material Aluminum Silicon slug, 6.35mm (0.25 in.) dia. x 6.35mm (0.25 in.) length

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'