missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Slug, 6.35mm (0.25 in.) dia x 6.35mm (0.25 in.) length, 99.99% (Metals basis), Thermo Scientific Chemicals

Form: Approx. 0.54g/slug

33.87 € - 285.42 €

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11344398
View Documents
Thermo Scientific Alfa Aesar
042327.09
10 g
33.87 €
10g
Please to purchase this item. Need a web account? Register with us today!
11364398
View Documents
Thermo Scientific Alfa Aesar
042327.18
50 g
77.28 €
50g
Please to purchase this item. Need a web account? Register with us today!
11354398
View Documents
Thermo Scientific Alfa Aesar
042327.30
250 g
285.42 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7429-90-5
26.98
XAGFODPZIPBFFR-UHFFFAOYSA-N
5359268
[Al]
Al
MFCD00134029
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
CHEBI:33629

Specifications

Silver
Silver-colored slug
Al
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
[Al]
26.98
CHEBI:33629
Slug
7429-90-5
99.99% (Metals basis)
MFCD00134029
XAGFODPZIPBFFR-UHFFFAOYSA-N
aluminium
5359268
Odorless
Aluminum
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-072-3

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.