Learn More
Cole-Parmer™ Aluminum Smooth Wall Weighing Dishes
Use these dishes for a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical and pharmaceutical. Cole-Parmer™ Aluminum Smooth Wall Weighing Dishes are ideal for storage, dispensing, general purpose sampling and testing.
Brand:  Cole-Parmer™ 01018-29
Product Code. 11735459
  • 223.00 € / Pack of 1000
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Smooth-sided, flat bottom, multipurpose “Utility” dishes
  • Oil-free and non-coated, no tab
  • Dish contour facilitates stacking and dispensing
  • Disposable
Specifications
Specifications
Flat
Single-use
Flat
For storage, dispensing, general purpose sampling and testing
1.7 cm
2.875 in.
80 mL
Aluminum
Weighing Dish
Weighing, Moisture Testing, Wastewater Analysis, Chemical, and Pharmaceutical
7.3 cm
1.7 cm