missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals

Product Code. 11311749
Click to view available options
Quantity:
500 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17927-65-0
Molecular Formula Al2O12S3
Molecular Weight (g/mol) 342.13
MDL Number MFCD00149136
InChI Key DIZPMCHEQGEION-UHFFFAOYSA-H
Synonym aluminum sulfate hydrate, dialuminum trisulfate hydrate, aluminum sulfate usp, aluminum sulfate 2:3 hydrate, alminum sulfate hydrate, aluminum sulfate usan, sulfuric acid, aluminum salt 3:2 , hydrate, 2al.3so4.h2o
PubChem CID 3080712
IUPAC Name dialuminium(3+) trisulfate
SMILES [Al+3].[Al+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

Color White
Physical Form Crystalline
Quantity 1 kg
Assay Percent Range >97%
Solubility Information It is slightly soluble in alcohol and dilute mineral acids.
Formula Weight 342.15 (Anhydrous)
Odor Odorless
Appearance White crystalline
Density 1.61
Chemical Name or Material Aluminum sulfate hydrate, hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'