missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum sulfate hydrate, 97+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
1 kg
5 kg
Description
Aluminum sulfate hydrate is a chemical compound commonly used as a flocculating agent in the purification of drinking water and waste water treatment plants, and also in paper manufacturing. Used as coagulant in decolorization and COD removal of secondary yeast wastewater effluents. It is also plays a significant role in arsenic removal by coagulation process.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | White |
Physical Form | Crystalline |
Quantity | 5 kg |
Assay Percent Range | >97% |
Solubility Information | It is slightly soluble in alcohol and dilute mineral acids. |
Formula Weight | 342.15 (Anhydrous) |
Odor | Odorless |
Appearance | White crystalline |
Density | 1.61 |
Chemical Name or Material | Aluminum sulfate hydrate, hydrate |
Safety and Handling
Product Identifier
- Aluminum sulfate hydrate
- Danger
- Serious eye damage/eye irritation Category 1
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Dialuminum trisulfate hydrate
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'