missing translation for 'onlineSavingsMsg'
Learn More
Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Honeywell Fluka™ Product Code.: 15611020

Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Honeywell Fluka™
BIOPHARMA

Product Code. 15611020 Shop All Honeywell Fluka Products
1 kg, 1kg
Click to view available options
Quantity:
1 kg
2.5 kg
20 kg
25 kg
6 x 1 kg
Unit Size:
1kg
2.5kg
20kg
25kg
Pack of 6
This item is not returnable. View return policy

Product Code. 15611020

Brand: Honeywell-Fluka 110441KG

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Puriss., meets analytical specification of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2H36O30S3
Molecular Weight (g/mol) 666.401
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18Show MoreShow Less
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Quantity 1 kg
Linear Formula Al2(SO4)3 · 18H2O
UN Number 3260
Formula Weight 342.15g/mol (anhydrous basis)
Percent Purity 100 to 110%
Grade puriss.
Chemical Name or Material Aluminum Sulfate Hydrate, Meets analytical specification of Ph.Eur.BP

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Product Code
Lot NumberA0397048 Certificate TypeCertificate of Analysis Date15/12/2022 Product Code110441000,110445000,110440050,11044-6X1KG,11044-1KG,11044-25KG
Lot NumberA0408899 Certificate TypeCertificate of Analysis Date13/12/2022 Product Code110441000,110445000,110440050,11044-6X1KG,11044-1KG,11044-25KG
Lot NumberA0440796 Certificate TypeCertificate of Analysis Date05/04/2022 Product Code
Lot NumberA0434862 Certificate TypeCertificate of Analysis Date02/10/2021 Product Code110441000,110445000,110440050,11044-6X1KG,11044-1KG,11044-25KG
Lot NumberA0419453 Certificate TypeCertificate of Analysis Date25/05/2021 Product Code110441000,110445000,110440050,11044-6X1KG,11044-1KG,11044-25KG
compliance-icons
Product Identifier
  • Aluminum sulfate hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Aluminum sulfate hydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum sulfate hydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 100-110%, 51.0-59.0% Al2(SO4)3 basis, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.