missing translation for 'onlineSavingsMsg'
Learn More

Aluminum sulfide, 99+%, (metals basis), Thermo Scientific Chemicals

240.20 € - 946.71 €

Chemical Identifiers

CAS 1302-81-4
Molecular Formula Al2S3
Molecular Weight (g/mol) 150.14
MDL Number MFCD00014162
InChI Key COOGPNLGKIHLSK-UHFFFAOYSA-N
IUPAC Name dialuminium(3+) trisulfanediide
SMILES [Al+3].[Al+3].[S--].[S--].[S--]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11357587
View Documents
Thermo Scientific Alfa Aesar
012851.06
5 g
240.20 €
5g
Please to purchase this item. Need a web account? Register with us today!
11347587
View Documents
Thermo Scientific Alfa Aesar
012851.14
25 g
946.71 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1302-81-4
150.14
COOGPNLGKIHLSK-UHFFFAOYSA-N
[Al+3].[Al+3].[S--].[S--].[S--]
Al2S3
MFCD00014162
dialuminium(3+) trisulfanediide

Specifications

1100°C
Yellow
>99% (Metal basis)
MFCD00014162
[Al+3].[Al+3].[S--].[S--].[S--]
150.14
Like rotten eggs
Yellowish granular
Aluminum sulfide
1302-81-4
Pieces
Al2S3
COOGPNLGKIHLSK-UHFFFAOYSA-N
dialuminium(3+) trisulfanediide
150.16
Moisture sensitive
2.02 g/cm3
Videos
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : DOT Class: 4.3, Packaging Group: III

missing translation for 'einecsNumber' : 215-109-0

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures; Store under Nitrogen

Product Identifier
  • Aluminum sulfide

Signal Word
  • Danger

Hazard Category
  • Substances or mixtures which\, in contact with water\, emits flammable gases Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only