missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Thermometer Stand

Supports 18 thermometers in an upright and secure position.

Brand:  Fisherbrand™ 6102

Product Code. 13078623

  • 44.95 € / Each
Estimated Shipment: 14-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • With protective foam cushion on the base
  • Manufactured from aluminum

Specifications

Specifications

18
Aluminum, Stainless Steel
Thermometer
120 mm
125 mm
120 mm
Thermometer Rack
18
1 Each
165 mm
125 mm
10 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.