Learn More
Cole-Parmer™ Aluminum Crimp-walled Weighing Dish
Use these weighing dishes for a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical and pharmaceutical. Aluminum Crimp-walled Weighing Dishes feature crimped sides for rigidity, sturdy gauge for heavy duty use.
Brand:  Cole-Parmer™ 01017-51
Product Code. 11779559
  • 132.00 € / Pack of 1000
Estimated Shipment: 06-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Use these weighing dishes for a wide variety of applications including weighing, moisture testing, wastewater analysis, chemical and pharmaceutical. Aluminum Crimp-walled Weighing Dishes feature crimped sides for rigidity, sturdy gauge for heavy duty use.
  • Dishes feature large tab for sample numbers
  • Flat bottom to improve sample handling
  • Sides are angled for easy separation
  • No oil residue, no vinyl coating to create contamination problems.
  • Smooth bottom
  • Crimpled walled with side finger tab handle 
  • Disposable
Specifications
Specifications
Flat
Single-use
Round
4.3 cm
1.3 cm
20 mL
Aluminum
Weighing Dish
1.75 in.