missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum wire, 0.5mm (0.02in) dia, annealed, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
Form: Approx. 0.53g/m; Temper: soft
59.40 € - 457.28 €
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11396837
|
Thermo Scientific Alfa Aesar
010748.G6 |
10 m |
59.40 €
10m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306847
|
Thermo Scientific Alfa Aesar
010748.H2 |
50 m |
212.06 €
50m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11316847
|
Thermo Scientific Alfa Aesar
010748.H7 |
250 m |
457.28 €
250m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
[Al] |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 |
Specifications
Silver | |
Silver-colored wire | |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Wire |
7429-90-5 | |
99.999% (Metals basis) | |
MFCD00134029 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminium | |
5359268 | |
Odorless | |
Aluminum, Annealed |
Safety and Handling
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only