missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 0.5mm (0.02in) dia, hard, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11309897
Click to view available options
Quantity:
10 m
250 m
50 m
This item is not returnable. View return policy
This item is not returnable. View return policy

Form: Approx.0.53g/m; Temper: as drawn

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
IUPAC Name aluminium
SMILES [Al]

Specifications

Color Silver
Appearance Silver-colored wire
Assay Percent Range 99.999% (Metals basis)
Quantity 50 m
Odor Odorless
Form Wire
Chemical Name or Material Aluminum, Hard

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'