missing translation for 'onlineSavingsMsg'
Learn More

Aluminum wire, 1.0mm (0.04in) dia, annealed, 99+% (metals basis), Thermo Scientific Chemicals

Product Code. 11311718
Click to view available options
Quantity:
10 m
250 m
50 m
This item is not returnable. View return policy
Normativa: Este producto contiene un precursor de explosivos de acuerdo con el Reglamento (CE) No 1148/2019, todos los clientes deben completar una declaración antes de la entrega.
Este artículo no se puede devolver. Vea la política de devoluciones
Normativa: Este producto contiene un precursor de explosivos de acuerdo con el Reglamento (CE) No 1148/2019, todos los clientes deben completar una declaración antes de la entrega.

Form: Approx. 2.1g/m; Temper: soft

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Especificaciones

Color Silver
Appearance Silver-colored wire
Assay Percent Range 99+% (Metals basis)
Quantity 10 m
IUPAC Name aluminum
Odor Odorless
Form Wire
Chemical Name or Material Aluminum, Annealed

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'