missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ AM 404

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Competitive and selective inhibitor of carrier-mediated anandamide transport (IC50 = 1 μM). Does not activate CB1 receptors or inhibit anandamide hydrolysis but has been shown to activate native and cloned vanilloid receptors (pEC50 = 7.4). Active in vivo.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 395.58 |
| Purity | >98% |
| Molecular Weight (g/mol) | 395.59 |
| Chemical Name or Material | AM 404 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction