Learn More
AM095 free acid, MedChemExpress
MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
Brand:  Medchem Express HY-16040
Encompass_Preferred
 View more versions of this product
Product Code. 16406978
  • 199.80 € / 10mg
Explore more special offers
Description
Description
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
Specifications
Specifications
AM095 free acid
Off-White
C27H24N2O5
DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming)
456.49
98.07%
Solid
1228690-36-5
Research
10 mg
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1
456.49
Research
COVID-19-immunoregulation