Learn More
AM095 free acid, MedChemExpress
MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
Brand: Medchem Express HY-16040
This item is not returnable.
View return policy
Description
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.Specifications
AM095 free acid | |
Off-White | |
C27H24N2O5 | |
DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming) | |
456.49 | |
98.07% | |
Solid |
1228690-36-5 | |
Research | |
200 mg | |
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1 | |
456.49 | |
Research | |
COVID-19-immunoregulation |