Learn More
AM095 free acid, MedChemExpress
MedChemExpress AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.
Brand: Medchem Express HY-16040
This item is not returnable.
View return policy
Description
AM095 (free acid) is a potent LPA1 receptor antagonist with IC50 values of 0.98 and 0.73 μM for recombinant human or mouse LPA1 respectively.Specifications
AM095 free acid | |
Off-White | |
C27H24N2O5 | |
DMSO : 67.3 mg/mL (147.43 mM; Need ultrasonic and warming) | |
456.49 | |
98.07% | |
Solid |
1228690-36-5 | |
Research | |
5 mg | |
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4)C)=O)C(C)=NO3)C=C2)C=C1 | |
456.49 | |
Research | |
COVID-19-immunoregulation |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
AM095 free acid, MedChemExpress