Learn More
AM211, MedChemExpress
MedChemExpress AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.
Brand: Medchem Express HY-13213
This item is not returnable.
View return policy
Description
AM211 is a potent, selective and orally bioavailable prostaglandin D2 (PGD2) receptor type 2 (DP2) antagonist, with IC50s of 4.9 nM, 7.8 nM, 4.9 nM, 10.4 nM for human, mouse, guinea pig, and rat DP2, respectively.Specifications
AM211 | |
White | |
C27H27F3N2O4 | |
AM211 free acid | |
O=C(O)CC1=CC(C2=CC=C(C(F)(F)F)C=C2CN(CC)C(NCC3=CC=CC=C3)=O)=C(OC)C=C1 | |
500.51 | |
Research | |
COVID-19-immunoregulation |
1175526-27-8 | |
Research | |
10 mg | |
DMSO : ≥ 125 mg/mL (249.75 mM) | |
500.51 | |
98.11% | |
Solid |