Learn More
AM966, MedChemExpress
MedChemExpress AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).
Brand: Medchem Express HY-15277
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
This item is not returnable.
View return policy
Description
AM966 is a high affinity, selective, oral LPA1-antagonist, inhibits LPA-stimulated intracellular calcium release (IC50=17 nM).Specifications
AM966 | |
Off-White | |
C27H23ClN2O5 | |
DMSO : 100 mg/mL (203.70 mM; Need ultrasonic) | |
490.93 | |
98.64% | |
Solid |
1228690-19-4 | |
Research | |
50 mg | |
O=C(O)CC1=CC=C(C2=CC=C(C3=C(NC(O[C@@H](C4=CC=CC=C4Cl)C)=O)C(C)=NO3)C=C2)C=C1 | |
490.93 | |
Research | |
COVID-19-immunoregulation |