missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers

Amber glass to protect light-sensitive contents.

Brand:  BRAND™ 37485

55.17 EUR valid until 2024-12-31
Use promo code "21985" to get your promotional price.


 View more versions of this product

Product Code. 15228816

  • 68.00 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Includes USP batch certificate
  • Class A, DE-M marking

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
50 mL
1
NS 12/21
A
11 mm
Borosilicate Glass (3.3)
Volumetric flask
±0.05 mL
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers