missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers

Amber glass to protect light-sensitive contents.

Brand:  BRAND™ 37490

 View more versions of this product

Product Code. 15258816

  • 124.00 € / Pack of 2
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Includes USP batch certificate
  • Class A, DE-M marking

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polypropylene
500 mL
1
NS 19/26
A
19 mm
Borosilicate Glass (3.3)
Volumetric flask
±0.20 mL
Stopper
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.