Learn More
BRAND™ Amber 3.3 Borosilicate Glass Volumetric Flask With Polypropylene Stoppers
Amber glass to protect light-sensitive contents.
Brand:  BRAND™ 37481
56.34 EUR valid until 2024-12-31
Use promo code "21985" to get your promotional price.
 View more versions of this product
Product Code. 15673967
  • 71.75 € / Pack of 2
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Manufactured from Boro 3.3 glass
  • Acc. to DIN EN ISO 1042
  • Includes USP batch certificate
  • Class A, DE-M marking
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PP (Polypropylene)
Borosilicate Glass (3.3)
A
5 mL
Volumetric flask
Stopper