missing translation for 'onlineSavingsMsg'
Learn More

AmberChrom 1x2 50-100 (Cl), Thermo Scientific Chemicals

Chemical Identifiers

CAS 9085-42-1
Molecular Formula C29H34ClN
Molecular Weight (g/mol) 432.05
MDL Number MFCD00132715
InChI Key BBQMUEOYPPPODD-UHFFFAOYSA-M
Synonym dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
PubChem CID 16212807
SMILES [Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11488990
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12130.18
50 g N/A
N/A
11498990
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12130.30
250 g N/A
N/A
11478990
View Documents Promotion Details
Thermo Scientific Alfa Aesar
A12130.0B
1000 g N/A
N/A
Description

Description

It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as a strongly basic (type I) anion exchange resin for food and drug processing chemical synthesis, medical research, mining, agriculture. It can be combined with any food product or ingested in any form.

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a tightly sealed container. Store away from oxidizing agents.
Specifications

Chemical Identifiers

9085-42-1
432.05
BBQMUEOYPPPODD-UHFFFAOYSA-M
16212807
C29H34ClN
MFCD00132715
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1

Specifications

9085-42-1
C29H34ClN
dowex 1x2 50-100 cl, dowex 1x8 50-100 cl
BBQMUEOYPPPODD-UHFFFAOYSA-M
1,4-diethenylbenzene 4-ethenyl-N,N,N-trimethylanilinium ethenylbenzene chloride
16212807
Amine-like
MFCD00132715
Insoluble in water.
[Cl-].C=CC1=CC=CC=C1.C=CC1=CC=C(C=C)C=C1.C[N+](C)(C)C1=CC=C(C=C)C=C1
432.05
Dowex™ 1X2 50-100 (Cl)
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only