missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ IRC-120(H), ion exchange resin, Thermo Scientific Chemicals

Product Code. 15439009
Click to view available options
Quantity:
250 g

Amberlite™ IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Amberlite|r IR-120(H) is a strongly acidic cation exchange resin which is used for a wide variety of water demineralization, dealkalization and deionization applications. It is also employed in the separation of amino acids at low pH.

Solubility
Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78922-04-0
Molecular Formula C13H10ClNO4S
Molecular Weight (g/mol) 311.736
MDL Number MFCD00132707
InChI Key APBOVLPLJFJSRI-UHFFFAOYSA-N
Synonym 3-3-chlorophenylsulfonamido benzoic acid, 3-3-chloro-benzenesulfonylamino-benzoic acid, 3-3-chlorophenyl sulfonyl amino benzoic acid, benzoicacid, 3-3-chlorophenyl sulfonyl amino, 3-3-chlorobenzenesulfonamido benzoic acid, amberlite ir-120, 3-3-chlorophenyl sulfonylamino benzoic acid, 3-3-chlorophenyl sulfonamido benzoic acid
PubChem CID 8190984
IUPAC Name 3-[(3-chlorophenyl)sulfonylamino]benzoic acid
SMILES C1=CC(=CC(=C1)NS(=O)(=O)C2=CC(=CC=C2)Cl)C(=O)O

Specifications

Density 1.28
Merck Index 14,382
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlite™ IR-120(H), ion exchange resin
compliance-icons
Product Identifier
  • Amberlite IR-120(H)
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Amberlite IR 120 Plus

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'