Learn More
Amberlite™ IRC120 Na ion exchange resin, Thermo Scientific Chemicals
Description
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Amberlite™ IR-120(Na) ion exchange resin is used in water softening, dealkalinization and deionization. It is used for the recovery of toxic metals. It is also used in separation medium for TLC, paper chromatography and LPLC.
Solubility
Insoluble in water.
Notes
Water softening, dealkalinization, deionization
Specifications
Specifications
Merck Index | 14,382 |
Quantity | 100 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Amberlite™ IR-120(Na) ion exchange resin |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.