missing translation for 'onlineSavingsMsg'
Learn More

Amberlite™ XAD-7 HP, Thermo Scientific Chemicals

35.07 € - 368.84 €

Chemical Identifiers

CAS 37380-43-1
MDL Number MFCD00132705
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10606062
SDS View Documents
Thermo Scientific Acros
202241000
100 g Plastic bottle
35.07 €
100g
Please to purchase this item. Need a web account? Register with us today!
10216260
SDS View Documents
Thermo Scientific Acros
202245000
500 g Plastic bottle
107.54 €
500g
Please to purchase this item. Need a web account? Register with us today!
11976991
SDS View Documents
Thermo Scientific Acros
202240025
2.5 kg Plastic bottle
368.84 €
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
Specifications

Chemical Identifiers

37380-43-1
MFCD00132705

Specifications

37380-43-1
MFCD00132705
Amberlite™ XAD-7 HP
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlite™ XAD-7 HP, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.