Learn More
Description
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Amberlite XAD-7 is used in chromatography media, enzyme immobilization and water purification. It is also used in the removal of relatively polar compounds from non aqueous solvents, non-aromatic compounds from polar solvent, recovery of plant extracts, antibiotics, enzymes and proteins.
Solubility
Insoluble in water.
Notes
Keep the container tightly sealed. Store in a cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.

Specifications
Specifications
Merck Index | 14,382 |
Quantity | 500 g |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Amberlite XAD-7 |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
A0456572 | Certificate of Analysis | 30/08/2024 | L19564.22,L19564.36 |
A0456571 | Certificate of Analysis | 22/04/2024 | L19564.22,L19564.36 |
A0450640 | Certificate of Analysis | 12/12/2023 | L19564.22,L19564,L19564.36 |
10238010 | Certificate of Analysis | 24/02/2023 | L19564.22,L19564.36 |
10231155 | Certificate of Analysis | 08/12/2021 | L19564.22,L19564.36 |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.