missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15(H), ion exchange resin

Product Code. 11369379
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11369379

Brand: Thermo Scientific Alfa Aesar 089079.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Strongly acidic macroreticular resin | CAS: 39389-20-3 | C18H18O3S | 314.399 g/mol

Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39389-20-3
Molecular Formula C18H18O3S
Molecular Weight (g/mol) 314.399
MDL Number MFCD00145841
InChI Key SIWVGXQOXWGJCI-UHFFFAOYSA-N
Synonym amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
PubChem CID 170197
IUPAC Name 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
SMILES C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O

Specifications

Physical Form Pale brown beads
Odor Odorless
Color Brown
Quantity 1 kg
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ 15(H)

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.