missing translation for 'onlineSavingsMsg'
Learn More

Amberlyst™ 15(H), wet, ion exchange resin, Thermo Scientific Chemicals

Product Code. 11345168
Click to view available options
Quantity:
250 g
1 kg
5 kg

Amberlyst™ 15(H) is a strongly acidic cation exchange resin used as a heterogeneous acid catalysis and is suitable for non-aqueous catalysis. It is used to determine the mechanism of phenol alkylation using density functional theory (DFT). It is widely utilized as a column packing material for HPLC analysis. It is employed in separation media for Thin-layer chromatography (TLC) and paper chromatography. It is used in various chemical reactions like esterification, transesterification, Michael addition, Prins cyclization, Friedel-Crafts alkylation, acylation, cleavage of epoxides and crossed-aldol condensation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 39389-20-3
Molecular Formula C18H18O3S
Molecular Weight (g/mol) 314.399
MDL Number MFCD00145841
InChI Key SIWVGXQOXWGJCI-UHFFFAOYSA-N
Synonym amberlyst 15, wet, ion exchange resin, 2-ethenylbenzenesulfonic acid; divinylbenzene, benzenesulfonic acid, ethenyl-, polymer with diethenylbenzene, ksc581g2r, amberlyst 15 ion-exchange resin, amberlite? ir120 hydrogen form, 2-ethenylbenzenesulfonic acid-1,2-diethenylbenzene 1:1, amberlite r ir120 hydrogen form, divinylbenzene-styrenesulfonic acid copolymer, 1,2-bis ethenyl benzene; 2-ethenylbenzenesulfonic acid
PubChem CID 170197
IUPAC Name 1,2-bis(ethenyl)benzene;2-ethenylbenzenesulfonic acid
SMILES C=CC1=CC=CC=C1C=C.C=CC1=CC=CC=C1S(=O)(=O)O

Specifications

Physical Form Beads
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Amberlyst™ 15 (H) ion exchange resin, Wet, Styrene-DVB; Strongly acidic macroreticular resin

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'