missing translation for 'onlineSavingsMsg'
Learn More
Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals Product Code.: 10052800

Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals

Product Code. 10052800
250 g, Plastic bottle
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10052800

Brand: Thermo Scientific Acros 202172500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Ion exchange resin
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9049-93-8
MDL Number MFCD00145842

Specifications

Physical Form Beads
Particle Size Particle size analysis: uniformity coefficient: <1.80, harmonic mean size: 0.490 to 0.690mm,% <0.300mm: <1.0%,%>1.180mm: <2.0%
Packaging Plastic bottle
Volume Total Capacity (free base):> 1.30 eq/L
Water Retention Capacity (free base): 52.0 to 62.0%
Fieser 06,302; 07,182; 08,263
Color Brown
Merck Index 10, 381
Quantity 250 g
Chemical Name or Material Amberlyst™ A-21
Product Identifier
  • Amberlyst]TS A-21 ion-exchange resin

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Amberlyst™ A-21, ion-exchange resin, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.