missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acid-Phenol:Chloroform, pH 4.5 (with IAA, 125:24:1)

In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing.

Brand:  Invitrogen™ AM9722

227.20 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 10544645

  • 284.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Acid Phenol:Chloroform:IAA (125:24:1) is premixed and supplied at pH 4.5 ± 0.2. Provided in one bottle of 400 mL. In RNA extraction procedures, Acid Phenol:Chloroform:IAA aids in the removal of DNA (it partitions into the organic phase), helps to stabilize the interface, and prevents foaming when mixing. Preparation of phenol for use in molecular biology applications is a time-consuming and often hazardous procedure due to its toxic and corrosive nature. Ambion premixed, quality-tested, saturated phenols are ready to use, eliminating handling problems and offering a more convenient, safer, and easier alternative to preparing solutions from crystalline phenol.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phenols
Bottle
Ambion™
400 mL
Liquid
Store at 4°C or -20°C.
4.5
Molecular Biology Grade
Room Temperature
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • ACID PHENOL:CHLOROFORM 400 ML

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 1

Hazard Statement
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H331-Toxic if inhaled.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : phenol, chloroform

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.