missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ammonium Acetate (5 M), RNase-free

Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use.

67.75 € - 160.00 €

Specifications

Chemical Name or Material Ammonium Acetate
Content And Storage Store at room temperature.
Concentration 5 M
Packaging Type Bottle
Product Line Ambion™
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10534645
View Documents
Invitrogen™
AM9070G
100 mL
67.75 €
Each
Please to purchase this item. Need a web account? Register with us today!
10075664
View Documents
Invitrogen™
AM9071
500 mL
160.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Invitrogen Molecular biology grade, 5 M Ammonium Acetate solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

Specifications

Specifications

Ammonium Acetate
5 M
Ambion™
Room Temperature
Store at room temperature.
Bottle
Molecular Biology Grade
Liquid
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.