missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ARCA (Anti-Reverse Cap Analog)
Cap analog for in vitro transcription for capped transcripts generation
Brand: Invitrogen™ AM8045
Includes: Single tube (10 units; 10 A255 U).
Description
Invitrogen™ Anti-Reverse Cap Analog (ARCA) is a cap analog used during in vitro transcription for the generation of capped transcripts. A modified cap analog in which the 3' OH group (closer to m7G) is replaced with –OCH3.
- Tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity
- Assayed for efficient synthesis of capped in vitro transcripts by using cap analog in functional MEGAscript™ In Vitro transcription and Retic Lysate translation reactions
Specifications
Supplied in a single tube to be stored at or below –70°C. Avoid multiple freeze-thaw cycles. Aliquots of the product may be stored short-term at –20°C. | |
Cap Analog for In vitro Transcription, Cap Analog for mRNA | |
mMessage mMAchine, MEGAscript | |
Ambion™ | |
10 Units |
40 mM | |
In Vitro Transcription, synthetic mRNA, mRNA | |
RNA Labeling | |
Cap analog for mRNA, cap analog for synthetic mRNA | |
Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.