missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR, compatible with either PAXgene™ or Tempus™ Blood RNA Tubes
GREENER_CHOICE

The Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR makes it possible to perform expression analysis directly from a 500 μL aliquot of human whole blood collected in PAXgene™ or Tempus™ Blood RNA tubes without conventional RNA purification.

Brand:  Invitrogen™ 4449079

Product Code. 11300922

  • 818.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: Kit Components: Box 1 - Digestion Solution 5mL, Tempus™ Pellet Enhancer 1mL, Tempus™ Wash No. 1 37.5mL, Tempus™ Wash No. 2 37.5mL, 2X PBS for Tempus™ 12.5mL, PAXgene™ Wash 75mL. Box 2 - Stop Solution 0.5, DNase 1, 50μL

Requires : Standard centrifuge capable of 5,000 x g with either 96-well plate or microfuge tube adaptors, 96-well plate or microfuge tube vortexer, blood tubes, reverse transcription reagents for cDNA synthesis, and TaqMan™ master mixes for real-time PCR analysis
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Kit Components:
Box 1-Store at 4°C:
• Digestion Solution, 5 mL
• Tempus™ Pellet Enhancer, 1 mL
• Tempus™ Wash #1, 37.5 mL
• Tempus™ Wash #2, 37.5 mL
• 2X PBS for Tempus™, 12.5 mL
• PAXgene™ Wash, 75 mL

Box 2-Store at -20 °C
• Stop Solution, 0.5 mL
• DNase 1, 50 μL

<1 hr.
Lysate
1-Step qRT-PCR, 2-Step qRT-PCR
50 Preps
50 preps
Tube, 96-well Plate
Blood
High-throughput Compatible, Not High-throughput Compatible (Manual), Automated Protocols
Fewer resources used and less waste, Less hazardous
Ambion™, Blood-to-CT™
Lysate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Stabilized Blood-to-CT™ Nucleic Acid Preparation Kit for qPCR, compatible with either PAXgene™ or Tempus™ Blood RNA Tubes > 50 preps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.