missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Buffer Kit, RNase-free

The Invitrogen Buffer Kit provides high quality, RNase free buffers for critical RNA analysis procedures.

Brand:  Invitrogen™ AM9010

286.40 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10722307

  • 358.00 € / Pack of 800

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Invitrogen Buffer Kit provides high quality, RNase free buffers for critical RNA analysis procedures. The kit consists of one 100 mL bottle of each of the following solutions: 1M Tris pH 7.0, 1M Tris pH 8.0, 5M NaCl, 1M MgCl2, 0.5M EDTA pH 8.0, 2M KCl, 5M Ammonium Acetate, and DEPC-treated H2O. Ammonium Acetate is sterile filtered. All other solutions are autoclaved post-packaging to ensure sterility. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Kit
Ambion™
1 set
miRNA analysis
Liquid
Buffer Kit
DNase-Free, RNase-Free
Room Temperature
8 x 100 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.