missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Cap Analog (m7G(5')ppp(5')G), 25 A254 Units
Click to view available options
Quantity:
1 Tube
Unit Size:
Each
Description
Includes
Single tube (25 A254 U).
Invitrogen Cap Analog [m7G(5')ppp(5')G] is used for the synthesis of 5' capped RNA molecules as part of in vitro transcription reactions. These molecules are translated more efficiently using in vitro translation systems than uncapped mRNAs. 25 A254 U supplied.
- Used as a highly specific inhibitor of the initiation step of protein synthesis.
- Tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity
- Assayed for efficient synthesis of capped in vitro transcripts by using cap analog in functional MEGAscript™ In Vitro transcription and Retic Lysate translation reactions
Specifications
Specifications
| Content And Storage | Supplied in a single tube to be stored at or below –70°C. Avoid multiple freeze-thaw cycles. Aliquots of the product may be stored short-term at –20°C. |
| Concentration | 40 mM |
| Final Product Type | Cap Analog for In vitro Transcription, Cap Analog for mRNA |
| For Use With (Application) | In Vitro Transcription, synthetic mRNA, mRNA |
| For Use With (Equipment) | mMessage mMAchine, MEGAscript |
| Labeling Method | RNA Labeling |
| Product Line | Ambion |
| Product Type | Cap analog for mRNA, cap analog for synthetic mRNA |
| Quantity | 1 Tube |
| Shipping Condition | Dry Ice |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction