missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ DEPC-Treated Water

Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC.

72.25 € - 306.00 €

Specifications

Grade Molecular Biology
Packaging Bottle
Treatment(s) DEPC-Treated
Purification Method Autoclaved, Membrane-Filtered
Product Line Ambion™
View More Specs

Products 6
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10289104
View Documents
Invitrogen™
AM9915G
1 x 100 mL
72.25 €
Each
Please to purchase this item. Need a web account? Register with us today!
10197484
View Documents
Invitrogen™
AM9920
1 x 500 mL
128.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
11597065
View Documents
Invitrogen™
AM9906
10 x 50 mL
216.00 €
Pack of 500
Please to purchase this item. Need a web account? Register with us today!
10565405
View Documents
Invitrogen™
AM9916
5 x 100 mL
199.00 €
500mL
Please to purchase this item. Need a web account? Register with us today!
10419224
View Documents
Invitrogen™
AM9922
1 x 1000 mL
177.00 €
Each
Please to purchase this item. Need a web account? Register with us today!
10514065
View Documents
Invitrogen™
4387937
4 x 1000 mL
306.00 €
4000mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Ambion DEPC-treated water is certified nuclease-free. DEPC-treated water is autoclaved pre- and post-packaging to ensure sterility and inactivation of DEPC. It is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Specifications

Specifications

Molecular Biology
DEPC-Treated
Ambion™
Bottle
Autoclaved, Membrane-Filtered
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at room temperature.

SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ DEPC-Treated Water

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.