missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ElectroZap™ Electrode Decontamination Solution

Invitrogen™ ElectroZap is a non-corrosive, stable cleaning solution that has been specially formulated to clean electrode surfaces in asimple, quick, and efficient manner and is effective against high levels of RNase or DNase contamination.

Brand:  Invitrogen™ AM9785

Product Code. 10458634

  • 144.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen™ ElectroZap is a non-corrosive, stable cleaning solution that has been specially formulated to clean electrode surfaces in asimple, quick, and efficient manner and is effective against high levels of RNase or DNase contamination. Researchers performing RNA analysis often require the use of pH meter to prepare their RNase-free solutions. The use of same pH meter by several laboratory personnel, however, can often lead to problems with RNase contamination, especially when preparing solutions that cannot be DEPC-treated or autoclaved. Special precautions must be taken when removing RNase contamination from pH meter so that the accuracy will be unaffected and there is no residue left by cleaning solution. Simply spray ElectroZap directly onto the electrode and then rinse off with RNase-free water — no residue that may interfere with subsequent enzymatic reactions remains. Supplied in 250 mL bottle.

  • Easily rinsed away leaving no residue
  • Stable, safe, and non-corrosive
  • No need for strong acid or alkali
  • Also removes DNases and other proteins

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Liquid
Room Temperature
RNase Control Reagent
RNA Extraction
Store ElectroZap™ at room temperature.
250 mL
Ambion™, ElectroZap
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2805908 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberAM9785
Lot Number2891649 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberAM9785
Lot Number3057309 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberAM9785
Lot Number2968736 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9785
Lot Number3008761 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberAM9785
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ ElectroZap™ Electrode Decontamination Solution > 250mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.