missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Elution Solution, Nucleic Acid Purification

The Elution Solution is used in the purification of total RNA from cells, primary cell isolates, animal and plant tissues and tumors.

Brand:  Invitrogen™ 4305893

184.50 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 15681752

  • 246.00 € / 1000mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Elution Solution is used in the purification of total RNA from cells, primary cell isolates, animal and plant tissues and tumors.

  • For use with Total RNA Lysis Solution and Nucleic Acid Purification
  • Size: 1000 mL

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature.
Nucleic Acid Purification
1000 mL
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.