Learn More
Invitrogen™ FirstChoice™ RLM-RACE Kit
Designed to amplify cDNA only from full-length, capped mRNA, usually producing a single band after PCR
Brand: Invitrogen™ AM1700
864.88 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Includes: Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate GAPDH-M, and 0.5M EDTA should be stored at –20°C. Nuclease-free water may be stored at any temperature.
Requires : SuperTaq Thermostable Taq DNA Polymerase sold separately. For optimal amplification of fragments ≥1 kb, use SuperTaq-Plus.
This item is not returnable. View return policy
Description
The FirstChoice RLM-RACE Kit is a major improvement over the basic rapid amplification of cDNA ends (RACE) protocol. The RLM-RACE procedure selects only full-length mRNA—no rRNA, tRNA or degraded RNA—and facilitates the cloning of sequences from the 5' ends of messages.
- From RNA to PCR product in less than a day
- Yields single, specific product from rare transcripts
- Selects 5' and/or 3' ends of true messages
- Efficient—
- Al enzymatic reactions are optimized to ensure detection of even rarest mRNA
- Final product size: 7kb or less
- Optimal reaction temperature: 42°C
- Sample: RNA
- Rapid amplification of cDNA ends (5'-RACE) is polymerase chain reaction-based technique developed to facilitate cloning of sequence from the 5'-ends of messages
- Improvement to basic RACE protocol
- Selects full-length mRNA—no rRNA, tRNA or degraded mRNA
Optimal reaction temperature: 42°C
Cloning, PCR and Real-Time PCR, Reverse Transcription, cDNA Libraries and Library Construction
Order Info
Shipping Condition: Dry ice
Specifications
RLM-RACE Kit | |
The following components should be stored at –20°C:
|
|
42°C | |
Dry Ice | |
Ambion™, FirstChoice™ | |
1 Kit |
Exo– Klenow, 10X Decamer Solution, 5X Reaction Buffer minus dATP, 5X Reaction Buffer minus dCTP, DECAtemplate GAPDH-M, and 0.5 M EDTA should be stored at -20°C. Nuclease-free water may be stored at any temperature. | |
cDNA | |
M-MLV | |
Kit | |
cDNA Libraries & Library Construction |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.