Learn More
Invitrogen™ MAXIscript™ SP6/T7 Transcription Kit
Fast synthesis of RNA probes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations
Brand:  Invitrogen™ AM1322
 View more versions of this product
Product Code. 10408644
  • 664.00 € / Each
Explore more special offers
Includes: SP6 Enzyme Mix, T7 Enzyme Mix, 10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, pTRI-b-actin, Nuclease-free Water
Description
Description
Ambion™ MAXIscript™ SP6/T7 In Vitro Transcription Kit synthesizes RNA probes in just 10 minutes for use in ribonuclease protection assays, in situ hybridization, and blot hybridizations. Each kit contains sufficient reagents for 15 reactions of each enzyme.
  • High yields of high specific activity, full-length RNA probes in just 10 minutes
  • Ideal probe synthesis kit for nuclease protection assays, Northern and Southern blots, and in situ hybridization
  • Includes TURBO DNase™
  • Contain quality-tested DNase/RNase-free reagents
  • Synthesizes RNA probes with specific activities reaching 1 x 109 cpm/μg in just 10 minutes
  • Effcient at synthesizing full-length probes, even at limiting nucleotide concentrations, making them the perfect companion to ribonuclease protection assays
  • For synthesis of nonisotopically labeled probes
  • Labeling can be accomplished post-transcriptionally by using Ambion's BrightStar™ Psoralen-Biotin Labeling Kit, or during transcription by incorporating modified nucleotides into reaction
  • Used for synthesis of modest amounts of unlabeled RNA (2–6μg/20μL reaction) of up to 2kb
TRUSTED_SUSTAINABILITY
Specifications
Specifications
SP6 Enzyme Mix, T7 Enzyme Mix,10X Transcription Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, TURBO DNase, Gel Loading Buffer II, and pTRI-b-actin are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Kit
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
Ambion™, MAXIscript™
100 Reactions
Dry Ice
PCR Products, Linearized DNA plasmid
Synthetic mRNA, mRNA
mRNA, Synthetic mRNA
100 Reactions
SP6/T7 In Vitro Transcription kit
T7, SP6
2-6 μg