missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAclear™ Transcription Clean-Up Kit

Product Code. 10600226
Click to view available options
Quantity:
20 Preps
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10600226

Brand: Invitrogen™ AM1908

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

The MEGAclear kit can be used to remove nucleotides, short oligonucleotides, proteins, and salts from RNA. The RNA recovered can be used for any application that requires high purity RNA. Recover from 1 ng to 500 μg of RNA.

The MEGAclear Transcription Clean-up Kit is designed for rapid high-throughput purification of RNA from enzymatic reactions such as in vitro transcription. The kit efficiently separates RNA from unincorporated NTPs, enzymes, and buffer components. The process is simple and fast, taking less than 30 minutes, and it recovers from 1 ng to 500 μg of RNA efficiently. The MEGAclear kit is appropriate for purification of ssRNA larger than 100 nt and dsRNA larger than 200 bp.

The MEGAclear kit purifies transcription reactions to produce high-quality RNA, free of unincorporated NTPs, enzymes, and buffer components, ideal for any downstream application that requires purified RNA. It can be used to remove nucleotides, short oligonucleotides,proteins, and salts from RNA. The RNA recovered can be used for any application that requires high purity RNA.

Use the MEGAclear lit to clean up any of the following:

  • In vitro transcribed RNA:
    • RNA from MEGAscript reactions
    • Amino allyl-modified RNA
    • Biotinyled RNA
    • Cy dye labeled RNA
    • Capped RNA (e.g., from mMESSAGE mMACHINE kit reactions)
  • Total RNA

Using the MEGAclear kit

The protocol is quick and easy. The transcription reaction is mixed with Binding Solution, passed over an RNA-binding filter, and washed; an elution step releases the purified RNA, which is now ready for any application requiring highly pure synthetic or in vitro transcribed RNA. For RNA recovery of 70% or greater, single-stranded RNA should be >100 nt and double-stranded RNA should be >200 bp.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Binding Solution, Wash Solution Concentrate, and 5 M Ammonium Acetate are stored at 4°C. Elution Solution can be stored at any temperature, and Filter Cartridges and Collection and Elution Tubes are stored at room temperature.
Final Product Type Purified Synthetic mRNA, Purified mRNA
Format Tube
Isolation Technology Silica Spin Column
Purification Target mRNA, Synthetic mRNA
Sample Type mRNA, Synthetic mRNA
For Use With (Application) In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
For Use With (Equipment) Vacuum Manifold, Centrifuge
High-throughput Compatibility Not High-throughput Compatible (Manual)
No. of Reactions 20 Preps
Product Line Ambion, MEGAclear
Product Type mRNA purification kit, Purification kit
Quantity 20 Preps
Purification Time 30 min
Shipping Condition Room Temperature
Starting Material Volume 1 ng to 500 μg of dsRNA>200b or ssRA>100bp
Yield 500 μg
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • MEGAclear Transcription Clean-Up Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Skin corrosion/irritation Category 1 C
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P330-Rinse mouth.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 2-Mercaptoethanol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.