missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MEGAscript™ T7 Transcription Kit

Ultra-high yield technology for in vitro transcription

Brand:  Invitrogen™ AMB13345

1639.10 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


 View more versions of this product

Product Code. 10600616

  • 2215.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Includes: T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II, Nuclease-free Water
This item is not returnable. View return policy

Description

Description

The high yields are achieved by modifying typical transcription reaction conditions so that very high nucleotide concentrations can be effectively used. The T7 Enzyme Mix and the 10X Reaction Buffer are specifically calibrated for each lot and RNA polymerase.

Using the MEGAscript T7 Kit:
The MEGAscript T7 Kit contains in vitro transcription reaction components and a control template. The kit will yield a total of 3 to 5mg of RNA (approximately 100μg of RNA or more per reaction) from the control template supplied with the kit. This corresponds to 400 to 650 moles of RNA for each mole of template. Smaller templates typically yield a lower mass and a higher molar yield of product. Novel transcription reaction conditions and a patented, high-yield technology allow the synthesis of 10 to 50 times the amount of RNA produced by conventional transcription reactions.

Applications:The MEGAscript T7 Kit is intended for the synthesis of large amounts of unlabeled or low specific activity RNA for a variety of uses, including in vitro translation, antisense/microinjection studies, and isolation of RNA binding proteins. In large-scale transcription reactions, the concentration of all four nucleotides is high, well above the Km for the enzyme. The MEGAscript T7 Kit typically yields over 10 times more RNA than conventional in vitro transcription reactions. The kit is not recommended for synthesis of high specific activity probes.

Accessory products:
MessageAmp™ aRNA amplification kits are powered by MEGAscript technology and have been developed especially for the amplification of cRNA prior to array analysis. To purify in vitro transcription products from free nucleotides, buffer components, and enzymes, use MEGAclear™ Kit (Mfr. No. AM1908) and the MEGAclear-96 Kit (Mfr.. No. AM1909).

RNA Yield:
100 μg

TRUSTED_SUSTAINABILITY
Specifications

Specifications

T7 Enzyme Mix, 10X Reaction Buffer, ATP Solution, CTP Solution, GTP Solution, UTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, and Gel Loading Buffer II are all stored at –20°C. Nuclease-free Water may be stored at any temperature.
Kit
In Vitro Transcription, synthetic mRNA, mRNA, transfection, LNPs, GENEart, linearized DNA plasmid, delivery, PCR products
Ambion™, MEGAscript™
200 reactions
Dry Ice
PCR Products, Linearized DNA plasmid
Synthetic mRNA, mRNA
mRNA, Synthetic mRNA
200 Reactions
T7 In Vitro Transcription kit
T7
100 μg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3095543 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberAMB13345
Lot Number3145373 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberAMB13345
Lot Number3145426 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberAMB13345
Lot Number3094288 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberAMB13345
Lot Number3094843 Certificate TypeCertificate of Analysis Date21/02/2025 Catalog NumberAMB13345
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ MEGAscript™ T7 Transcription Kit > 200 reactions

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.