missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NorthernMax™ Prehybridization/Hybridization Buffer

No BSA, Denhardt's, or other potentially nuclease-contaminated biological materials are used in preparing this buffer, ensuring maximum sensitivity and reproducibility

Brand:  Invitrogen™ AM8677

279.00 EUR valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10422655

  • 465.00 € / Each
Estimated Shipment: 18-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Functionally tested by northern blotting and is certified Rnase-free
  • Contains 50% formamide with proprietary blocking and background reduction agents
  • Blocking agents minimize cross hybridization to ribosomal sequences
  • Signal-to-noise ratios are increased by the background reducing agents' suppression of hybridization to the membrane, which is particularly important with nonisotopic detection protocols
  • Same component included in the NorthernMax Ultrasensitive Northern Blotting Kits but in a larger size
  • Ready to use
  • Comprehensive instructions are included

DNA and RNA Purification and Analysis, Northern Blotting, Nucleic Acid Gel Electrophoresis and Blotting

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at 4°C.
Northern Analysis Prehybridization/Hybridization Buffer
Room Temperature
10X
500 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.