missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Nuclease-Free Water (not DEPC-Treated)

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving.

Brand:  Invitrogen™ AM9937

 View more versions of this product

Product Code. 10526945

  • 208.00 € / 500mL
In Stock
to see stock.
Explore more special offers
Add to basket

Includes: 10 x 50 mL bottles.
This item is not returnable. View return policy

Description

Description

Nuclease-free Water (not DEPC-treated) has been deionized, filtered into the final bottle, and autoclaved. It is ready to use and requires no preparation, mixing, or autoclaving. This nuclease-free water is rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Using Nuclease-free Water
Because this water is not DEPC-treated, it is recommended for applications for which residual DEPC might prove detrimental (e.g., oocyte microinjection).

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

6 to 8
Bottle
10 x 50 mL
Room Temperature
Molecular Biology
Not DEPC-Treated
Autoclaved, Membrane-Filtered
Ambion™
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.