missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Potassium Acetate (3 M), pH 5.5, RNase-free

Invitrogen Molecular biology grade, 3M Potassium Acetate, pH 5.5 solution is supplied in one bottle containing 100 mL.

Brand:  Invitrogen™ AM9610

52.60 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10187824

  • 65.75 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Invitrogen Molecular biology grade, 3M Potassium Acetate, pH 5.5 solution is supplied in one bottle containing 100 mL. The solution is certified RNase-free, economical, and ready-to-use. Due to the ubiquitous presence of RNases, manufacturing products for use with RNA is especially challenging. Invitrogen's nuclease-free reagents and buffers are manufactured in facilities specifically designed to prevent the introduction of nucleases. Highly sensitive RNase assays are performed at several different stages of the manufacturing process to ensure the highest quality. These reagents are rigorously tested for contaminating nonspecific endonuclease, exonuclease, and RNase activity.

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Potassium Acetate
3 M
Bottle
Ambion™
100 mL
Liquid
Store at room temperature.
RNA Applications
5.5
Molecular Biology Grade
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures.